Search
Search

MED13L| Gene Symbol: MED13L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647096.1 | XP_023502864.1 | 2125 | 89 | ||
| XM_023647097.1 | XP_023502865.1 | 2125 | 89 | |||
| XM_023647098.1 | XP_023502866.1 | 1724 | 89 | |||
| XM_023647099.1 | XP_023502867.1 | 1890 | 89 | |||
| XM_023647100.1 | XP_023502868.1 | 3294 | 89 | |||