Search
Search
| Gene Symbol: TBX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005612582.3 | XP_005612639.1 | 1162 | 119 | ||
| XM_005612583.3 | XP_005612640.1 | 1241 | 119 | |||
| XM_023647104.1 | XP_023502872.1 | 1619 | 119 | |||
| XM_023647106.1 | XP_023502874.1 | 2031 | 119 | |||
Set Membership: |
|