Search
Search
| Gene Symbol: CHFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023648160.1 | XP_023503928.1 | 751 | 101 | ||
| XM_023648161.1 | XP_023503929.1 | 691 | 101 | |||
| XM_023648162.1 | XP_023503930.1 | 751 | 101 | |||
| XM_023648163.1 | XP_023503931.1 | 751 | 101 | |||
Set Membership: |
|