Search
Search

SEH1L| Gene Symbol: SEH1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647481.1 | XP_023503249.1 | 620 | 97 | ||
| XM_023647482.1 | XP_023503250.1 | 620 | 97 | |||
| GenBank mRNA | JL628759.1 | - | 1 - 2 | 116 | 97 | |
Set Membership: |
|