Search
Search

RBBP8| Gene Symbol: RBBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001915082.5 | XP_001915117.5 | 2880 | 72 | ||
| XM_023647501.1 | XP_023503269.1 | 3545 | 72 | |||
| XM_023647502.1 | XP_023503270.1 | 3549 | 72 | |||
| XM_023647503.1 | XP_023503271.1 | 3527 | 72 | |||
| XM_023647504.1 | XP_023503272.1 | 3440 | 72 | |||
| GenBank mRNA | JL621598.1 | - | 8 - 9 | 1692 | 72 | |
Set Membership: |
|