Search
Search
| Gene Symbol: EPG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498555.5 | XP_001498605.3 | 4069 | 112 | ||
| XM_014727678.2 | XP_014583164.2 | 4069 | 112 | |||
| XM_014727679.2 | XP_014583165.2 | 4069 | 112 | |||
Set Membership: |
|