Search
Search

NCOA2| Gene Symbol: NCOA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005613069.3 | XP_005613126.1 | 4333 | 118 | ||
| XM_005613070.3 | XP_005613127.1 | 4238 | 118 | |||
| XM_014727956.2 | XP_014583442.1 | 4384 | 118 | |||
| XM_023648481.1 | XP_023504249.1 | 4289 | 118 | |||
| XM_023648482.1 | XP_023504250.1 | 4278 | 118 | |||
| XM_023648483.1 | XP_023504251.1 | 4764 | 118 | |||
| XM_023648484.1 | XP_023504252.1 | 4082 | 118 | |||
Set Membership: |
|