Search
Search

CHCHD7| Gene Symbol: CHCHD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023648583.1 | XP_023504351.1 | 290 | 73 | ||
| XM_023648584.1 | XP_023504352.1 | 394 | 73 | |||
| XM_023648585.1 | XP_023504353.1 | 399 | 73 | |||
| XM_023648586.1 | XP_023504354.1 | 211 | 73 | |||
| XM_023648587.1 | XP_023504355.1 | 91 | 73 | |||
| XM_023648588.1 | XP_023504356.1 | 240 | 73 | |||