Search
Search

DECR1| Gene Symbol: DECR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488668.4 | XP_001488718.2 | 474 | 104 | ||
| XM_005613191.3 | XP_005613248.1 | 542 | 104 | |||
| XM_023648666.1 | XP_023504434.1 | 435 | 104 | |||
Set Membership: |
|