Search
Search

RGS22| Gene Symbol: RGS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023648761.1 | XP_023504529.1 | 6048 | 74 | ||
| XM_023648762.1 | XP_023504530.1 | 5265 | 74 | |||
| XM_023648763.1 | XP_023504531.1 | 6048 | 74 | |||
| XM_023648764.1 | XP_023504532.1 | 6023 | 74 | |||
| XM_023648765.1 | XP_023504533.1 | 5926 | 74 | |||
| XM_023648766.1 | XP_023504534.1 | 5863 | 74 | |||