Search
Search

ASAP1| Gene Symbol: ASAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023648975.1 | XP_023504743.1 | 1057 | 100 | ||
| XM_023648976.1 | XP_023504744.1 | 1047 | 100 | |||
| XM_023648977.1 | XP_023504745.1 | 1057 | 100 | |||