Search
Search

DENND3| Gene Symbol: DENND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005613316.3 | XP_005613373.1 | 3358 | 81 | ||
| XM_023649021.1 | XP_023504789.1 | 3175 | 81 | |||
| XM_023649022.1 | XP_023504790.1 | 3175 | 81 | |||