Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ec07007910_g1
See other PLEC GE Assays ›
Gene Symbol:
PLEC
Gene Name:
plectin
Gene Aliases:
-
Chromosome Location:
Chr.9: 84697562 - 84754010 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID
Ec07007910_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: PLEC
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_023649156.1
XP_023504924.1
3569
80
XM_023649157.1
XP_023504925.1
3554
80
XM_023649158.1
XP_023504926.1
3532
80
XM_023649159.1
XP_023504927.1
3517
80
XM_023649160.1
XP_023504928.1
4064
80
XM_023649161.1
XP_023504929.1
4049
80
XM_023649162.1
XP_023504930.1
4028
80
XM_023649163.1
XP_023504931.1
4012
80
XM_023649164.1
XP_023504932.1
3594
80
XM_023649165.1
XP_023504933.1
3579
80
XM_023649166.1
XP_023504934.1
3696
80
XM_023649167.1
XP_023504935.1
3669
80
XM_023649168.1
XP_023504936.1
3680
80
XM_023649169.1
XP_023504937.1
3399
80
XM_023649170.1
XP_023504938.1
3410
80
XM_023649171.1
XP_023504939.1
3644
80
XM_023649172.1
XP_023504940.1
3590
80
XM_023649174.1
XP_023504942.1
3629
80
XM_023649175.1
XP_023504943.1
3575
80
XM_023649176.1
XP_023504944.1
3614
80
XM_023649177.1
XP_023504945.1
3465
80
XM_023649178.1
XP_023504946.1
3593
80
XM_023649179.1
XP_023504947.1
3450
80
XM_023649180.1
XP_023504948.1
3539
80
XM_023649181.1
XP_023504949.1
3577
80
XM_023649182.1
XP_023504950.1
3414
80
XM_023649183.1
XP_023504951.1
3483
80
XM_023649185.1
XP_023504953.1
3468
80
XM_023649186.1
XP_023504954.1
3432
80
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Equus caballus
Panther Classification:
Molecular Function -
intermediate filament binding protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline