Search
Search

DGAT1| Gene Symbol: DGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005613365.3 | XP_005613422.2 | 915 | 84 | ||
| XM_023649292.1 | XP_023505060.1 | 974 | 84 | |||
| XM_023649293.1 | XP_023505061.1 | 828 | 84 | |||
Set Membership: |
|