Search
Search
| Gene Symbol: AGRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023636081.1 | XP_023491849.1 | 5684 | 82 | ||
| XM_023636082.1 | XP_023491850.1 | 5672 | 82 | |||
| XM_023636083.1 | XP_023491851.1 | 5666 | 82 | |||
| XM_023636084.1 | XP_023491852.1 | 5684 | 82 | |||
| XM_023636085.1 | XP_023491853.1 | 5653 | 82 | |||
| XM_023636086.1 | XP_023491854.1 | 5489 | 82 | |||
| XM_023636087.1 | XP_023491855.1 | 5433 | 82 | |||
| XM_023636088.1 | XP_023491856.1 | 5406 | 82 | |||
Set Membership: |
|