Search
Search

RBM33| Gene Symbol: RBM33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023640066.1 | XP_023495834.1 | 2561 | 109 | ||
| XM_023640067.1 | XP_023495835.1 | 2561 | 109 | |||
| XM_023640068.1 | XP_023495836.1 | 2561 | 109 | |||
Set Membership: |
|