Search
Search
| Gene Symbol: MSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014728759.1 | XP_014584245.1 | 547 | 114 | ||
| XM_023634415.1 | XP_023490183.1 | 536 | 114 | |||
| XM_023634416.1 | XP_023490184.1 | 479 | 114 | |||
| XM_023634417.1 | XP_023490185.1 | 537 | 114 | |||
Set Membership: |
|