Search
Search
| Gene Symbol: OTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488750.6 | XP_001488800.3 | 532 | 107 | ||
| XM_023633819.1 | XP_023489587.1 | 399 | 107 | |||
| XM_023633820.1 | XP_023489588.1 | 562 | 107 | |||
| XM_023633821.1 | XP_023489589.1 | 434 | 107 | |||
Set Membership: |
|