Search
Search

HDAC6| Gene Symbol: HDAC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614119.3 | XP_005614176.2 | 1007 | 82 | ||
| XM_005614121.3 | XP_005614178.2 | 974 | 82 | |||
| XM_005614122.3 | XP_005614179.2 | 902 | 82 | |||
| XM_005614123.2 | XP_005614180.2 | 892 | 82 | |||
| XM_005614126.3 | XP_005614183.2 | 1056 | 82 | |||
| XM_005614127.3 | XP_005614184.2 | 755 | 82 | |||
| XM_005614128.3 | XP_005614185.2 | 1010 | 82 | |||
| XM_005614129.3 | XP_005614186.2 | 605 | 82 | |||
| XM_014728836.2 | XP_014584322.2 | 932 | 82 | |||
| XM_014728837.2 | XP_014584323.2 | 984 | 82 | |||
| XM_014728838.2 | XP_014584324.2 | 878 | 82 | |||
| XM_023634476.1 | XP_023490244.1 | 782 | 82 | |||
| XM_023634477.1 | XP_023490245.1 | 670 | 82 | |||
| XM_023634478.1 | XP_023490246.1 | 667 | 82 | |||