Search
Search

DIAPH2| Gene Symbol: DIAPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001914669.5 | XP_001914704.3 | 1663 | 93 | ||
| XM_023634104.1 | XP_023489872.1 | 1684 | 93 | |||
| XM_023634105.1 | XP_023489873.1 | 1623 | 93 | |||
| XM_023634106.1 | XP_023489874.1 | 1651 | 93 | |||
| XM_023634107.1 | XP_023489875.1 | 1684 | 93 | |||