Search
Search

DIAPH2| Gene Symbol: DIAPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001914669.5 | XP_001914704.3 | 3040 | 100 | ||
| XM_023634104.1 | XP_023489872.1 | 3061 | 100 | |||
| XM_023634105.1 | XP_023489873.1 | 3000 | 100 | |||
| XM_023634106.1 | XP_023489874.1 | 3028 | 100 | |||
| XM_023634107.1 | XP_023489875.1 | 3061 | 100 | |||
| GenBank mRNA | JL638522.1 | - | 5 - 6 | 645 | 100 | |
Set Membership: |
|