Search
Search

TAF7L| Gene Symbol: TAF7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729047.2 | XP_014584533.1 | 1346 | 90 | ||
| XM_023633563.1 | XP_023489331.1 | 1405 | 90 | |||
| XM_023633564.1 | XP_023489332.1 | 1403 | 90 | |||
| XM_023633565.1 | XP_023489333.1 | 1344 | 90 | |||
Set Membership: |
|