Search
Search

COL4A6| Gene Symbol: COL4A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729082.2 | XP_014584568.2 | 3449 | 58 | ||
| XM_023633922.1 | XP_023489690.1 | 3395 | 58 | |||
| XM_023633923.1 | XP_023489691.1 | 3449 | 58 | |||
| XM_023633924.1 | XP_023489692.1 | 3395 | 58 | |||
| XM_023633925.1 | XP_023489693.1 | 3370 | 58 | |||
| XM_023633926.1 | XP_023489694.1 | 3278 | 58 | |||
Set Membership: |
|