Search
Search
| Gene Symbol: OCRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023634242.1 | XP_023490010.1 | 665 | 142 | ||
| XM_023634243.1 | XP_023490011.1 | 666 | 142 | |||
| XM_023634244.1 | XP_023490012.1 | 458 | 142 | |||
| XM_023634245.1 | XP_023490013.1 | 337 | 142 | |||
Set Membership: |
|