Search
Search
| Gene Symbol: F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498904.4 | XP_001498954.2 | 594 | 64 | ||
| XM_005614666.3 | XP_005614723.2 | 594 | 64 | |||
| XM_014729308.2 | XP_014584794.2 | 594 | 64 | |||
| XM_014729309.2 | XP_014584795.2 | 594 | 64 | |||
| XM_014729310.2 | XP_014584796.2 | 594 | 64 | |||
| XM_014729311.2 | XP_014584797.2 | 595 | 64 | |||
| XM_014729312.2 | XP_014584798.2 | 595 | 64 | |||
| XM_023634329.1 | XP_023490097.1 | 595 | 64 | |||
Set Membership: |
|