Search
Search

BRCC3| Gene Symbol: BRCC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498791.6 | XP_001498841.2 | 203 | 98 | ||
| XM_003365914.4 | XP_003365962.1 | 200 | 98 | |||
| GenBank mRNA | JL615923.1 | - | 1 - 2 | 135 | 98 | |
Set Membership: |
|