Search
Search

BRCC3| Gene Symbol: BRCC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498791.6 | XP_001498841.2 | 576 | 102 | ||
| XM_003365914.4 | XP_003365962.1 | 573 | 102 | |||
| XM_023634086.1 | XP_023489854.1 | 524 | 102 | |||
| XM_023634087.1 | XP_023489855.1 | 506 | 102 | |||
| GenBank mRNA | JL615923.1 | - | 6 - 7 | 508 | 102 | |
Set Membership: |
|