Search
Search

OSBPL1A| Gene Symbol: OSBPL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014727654.2 | XP_014583140.1 | 2216 | 86 | ||
| XM_023647580.1 | XP_023503348.1 | 1691 | 86 | |||
| XM_023647581.1 | XP_023503349.1 | 1318 | 86 | |||
| GenBank mRNA | JL618673.1 | - | 1 - 2 | 94 | 86 | |
Set Membership: |
|