Search
Search

SENP3| Gene Symbol: SENP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005597753.3 | XP_005597810.1 | 1741 | 99 | ||
| XM_005597754.3 | XP_005597811.1 | 1741 | 99 | |||
| GenBank mRNA | JL632171.1 | - | 1 - 2 | 129 | 99 | |
Set Membership: |
|