Search
Search

LOC106783094| Gene Symbol: LOC106783094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023639856.1 | XP_023495624.1 | 705 | 61 | ||
| GenBank mRNA | AY921369.1 | - | 1 - 2 | 73 | 61 | |
Set Membership: |
|