Search
Search

MFSD1| Gene Symbol: MFSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023623228.1 | XP_023478996.1 | 1365 | 112 | ||
| XM_023623230.1 | XP_023478998.1 | 1408 | 112 | |||
| XM_023623231.1 | XP_023478999.1 | 1200 | 112 | |||
| GenBank mRNA | JL616844.1 | - | 3 - 4 | 260 | 112 | |
Set Membership: |
|