Search
Search

MAGI1| Gene Symbol: MAGI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001487929.5 | XP_001487979.3 | 673 | 101 | ||
| XM_023621184.1 | XP_023476952.1 | 673 | 101 | |||
| XM_023621185.1 | XP_023476953.1 | 673 | 101 | |||
| XM_023621186.1 | XP_023476954.1 | 673 | 101 | |||
| XM_023621187.1 | XP_023476955.1 | 673 | 101 | |||
| XM_023621188.1 | XP_023476956.1 | 673 | 101 | |||
| XM_023621189.1 | XP_023476957.1 | 673 | 101 | |||
| XM_023621190.1 | XP_023476958.1 | 673 | 101 | |||
| XM_023621191.1 | XP_023476959.1 | 673 | 101 | |||
| XM_023621192.1 | XP_023476960.1 | 673 | 101 | |||
| XM_023621193.1 | XP_023476961.1 | 673 | 101 | |||
| XM_023621194.1 | XP_023476962.1 | 674 | 101 | |||
| XM_023621195.1 | XP_023476963.1 | 674 | 101 | |||
| XM_023621196.1 | XP_023476964.1 | 674 | 101 | |||
| XM_023621197.1 | XP_023476965.1 | 674 | 101 | |||
| XM_023621198.1 | XP_023476966.1 | 674 | 101 | |||
| XM_023621199.1 | XP_023476967.1 | 673 | 101 | |||
Set Membership: |
|