Search
Search

FMNL2| Gene Symbol: FMNL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023622915.1 | XP_023478683.1 | 455 | 107 | ||
| XM_023622916.1 | XP_023478684.1 | 455 | 107 | |||
| XM_023622917.1 | XP_023478685.1 | 455 | 107 | |||
| XM_023622918.1 | XP_023478686.1 | 455 | 107 | |||
| XM_023622923.1 | XP_023478691.1 | 455 | 107 | |||
Set Membership: |
|