Search
Search
| Gene Symbol: GRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014738217.2 | XP_014593703.2 | 3242 | 69 | ||
| XM_023633059.1 | XP_023488827.1 | 3419 | 69 | |||
| XM_023633060.1 | XP_023488828.1 | 3512 | 69 | |||
Set Membership: |
|