Search
Search

NLRP14| Gene Symbol: NLRP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645944.1 | XP_023501712.1 | 11025 | 119 | ||
| XM_023645945.1 | XP_023501713.1 | 7251 | 119 | |||
| XM_023645946.1 | XP_023501714.1 | 12473 | 119 | |||
| XM_023645947.1 | XP_023501715.1 | 12473 | 119 | |||
| XM_023645948.1 | XP_023501716.1 | 12473 | 119 | |||
Set Membership: |
|