Search
Search

LOC100072605| Gene Symbol: LOC100072605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614294.3 | XP_005614351.1 | 3346 | 80 | ||
| XM_005614295.3 | XP_005614352.1 | 3439 | 80 | |||
Set Membership: |
|