Search
Search
| Gene Symbol: PRX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023649758.1 | XP_023505526.1 | 1974 | 68 | ||
| XM_023649759.1 | XP_023505527.1 | 2638 | 68 | |||
| XM_023649760.1 | XP_023505528.1 | 1976 | 68 | |||
| XM_023649761.1 | XP_023505529.1 | 2634 | 68 | |||
| XM_023649763.1 | XP_023505531.1 | 1875 | 68 | |||
| XM_023649764.1 | XP_023505532.1 | 2539 | 68 | |||
Set Membership: |
|