Search
Search

DCLK2| Gene Symbol: DCLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001915409.5 | XP_001915444.3 | 2491 | 83 | ||
| XM_023627737.1 | XP_023483505.1 | 2488 | 83 | |||
| XM_023627745.1 | XP_023483513.1 | 2438 | 83 | |||
| XM_023627754.1 | XP_023483522.1 | 2435 | 83 | |||
Set Membership: |
|