Search
Search

LOC100067994| Gene Symbol: LOC100067994 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501871.4 | XP_001501921.3 | 1184 | 81 | ||
| XM_023638020.1 | XP_023493788.1 | 1141 | 81 | |||
Set Membership: |
|