Search
Search
| Gene Symbol: BCAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500363.6 | XP_001500413.3 | 1288 | 84 | ||
| XM_014739955.2 | XP_014595441.1 | 1255 | 84 | |||
| XM_014739956.2 | XP_014595442.1 | 2344 | 84 | |||
| XM_023640986.1 | XP_023496754.1 | 1284 | 84 | |||
| XM_023640987.1 | XP_023496755.1 | 1288 | 84 | |||
Set Membership: |
|