Search
Search

MSTO1| Gene Symbol: MSTO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494815.4 | XP_001494865.1 | 1538 | 86 | ||
| XM_005610031.3 | XP_005610088.1 | 1550 | 86 | |||
| XM_023641036.1 | XP_023496804.1 | 1542 | 86 | |||