Search
Search

CRTC2| Gene Symbol: CRTC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001495407.5 | XP_001495457.1 | 859 | 110 | ||
| XM_023641112.1 | XP_023496880.1 | 866 | 110 | |||
| XM_023641113.1 | XP_023496881.1 | 863 | 110 | |||
Set Membership: |
|