Search
Search
| Gene Symbol: RFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014738631.2 | XP_014594117.1 | 478 | 83 | ||
| XM_014738632.2 | XP_014594118.1 | 478 | 83 | |||
| XM_014738634.2 | XP_014594120.1 | 547 | 83 | |||
| XM_023638219.1 | XP_023493987.1 | 478 | 83 | |||
| GenBank mRNA | JL617868.1 | - | 1 - 2 | 134 | 83 | |
Set Membership: |
|