Search
Search

ZRANB1| Gene Symbol: ZRANB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489580.6 | XP_001489630.2 | 1722 | 100 | ||
| XM_014733167.2 | XP_014588653.1 | 986 | 100 | |||
| XM_023637362.1 | XP_023493130.1 | 2006 | 100 | |||
| XM_023637369.1 | XP_023493137.1 | 1875 | 100 | |||
Set Membership: |
|