Search
Search

CFAP43| Gene Symbol: CFAP43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014733245.2 | XP_014588731.2 | 3220 | 112 | ||
| XM_014733246.2 | XP_014588732.2 | 3144 | 112 | |||
| XM_023639855.1 | XP_023495623.1 | 3241 | 112 | |||
| XM_023639858.1 | XP_023495626.1 | 3217 | 112 | |||
| XM_023639860.1 | XP_023495628.1 | 3070 | 112 | |||
| XM_023639861.1 | XP_023495629.1 | 3003 | 112 | |||
| XM_023639863.1 | XP_023495631.1 | 3131 | 112 | |||
| XM_023639867.1 | XP_023495635.1 | 3220 | 112 | |||
| XM_023639869.1 | XP_023495637.1 | 2785 | 112 | |||
Set Membership: |
|