Search
Search

ARMH3| Gene Symbol: ARMH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499750.5 | XP_001499800.3 | 1167 | 83 | ||
| XM_023640534.1 | XP_023496302.1 | 1074 | 83 | |||
| XM_023640543.1 | XP_023496311.1 | 1171 | 83 | |||
| XM_023640545.1 | XP_023496313.1 | 850 | 83 | |||
| XM_023640550.1 | XP_023496318.1 | 869 | 83 | |||
| XM_023640560.1 | XP_023496328.1 | 832 | 83 | |||
| XM_023640562.1 | XP_023496330.1 | 821 | 83 | |||
| XM_023640569.1 | XP_023496337.1 | 1173 | 83 | |||