Search
Search

PDZD7| Gene Symbol: PDZD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023640751.1 | XP_023496519.1 | 1235 | 100 | ||
| XM_023640753.1 | XP_023496521.1 | 1235 | 100 | |||
| XM_023640757.1 | XP_023496525.1 | 1235 | 100 | |||
Set Membership: |
|