Search
Search

LZTS2| Gene Symbol: LZTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005602288.3 | XP_005602345.1 | 1834 | 82 | ||
| XM_005602289.3 | XP_005602346.1 | 1742 | 82 | |||
| XM_005602290.3 | XP_005602347.1 | 2142 | 82 | |||
| XM_014733071.2 | XP_014588557.1 | 2390 | 82 | |||
| XM_023640787.1 | XP_023496555.1 | 1771 | 82 | |||
Set Membership: |
|