Search
Search

CEP55| Gene Symbol: CEP55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005602360.3 | XP_005602417.1 | 1399 | 100 | ||
| XM_014733430.2 | XP_014588916.1 | 1399 | 100 | |||
| XM_023642416.1 | XP_023498184.1 | 1629 | 100 | |||
| XM_023642419.1 | XP_023498187.1 | 1179 | 100 | |||
| GenBank mRNA | JL615962.2 | - | 6 - 7 | 909 | 100 | |
Set Membership: |
|