Search
Search

STOX1| Gene Symbol: STOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023644104.1 | XP_023499872.1 | 2946 | 144 | ||
| XM_023644105.1 | XP_023499873.1 | 2888 | 144 | |||
| XM_023644106.1 | XP_023499874.1 | 2913 | 144 | |||
| XM_023644107.1 | XP_023499875.1 | 3897 | 144 | |||
| GenBank mRNA | JL630765.1 | - | 1 - 2 | 186 | 144 | |
Set Membership: |
|